Imprimer
Bioanalyzer Protein Analysis
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5067-1515 5067-1516 5067-1517 5067-1518
AGLS5067-1515EA 1380 EUR
AGLS5067-1515 AGLS5067-1516 AGLS5067-1517 AGLS5067-1518
Bioanalyzer Protein Analysis
Systèmes d'électrophorèse Capillaires pour électrophorèse
Electrophoretic separation of proteins and peptides up to 230 kD.
  • Flexibility to choose the appropriate kit based on the sizing range
  • Minimal sample consumption is required, with only 4 µl of sample needed for an accurate analysis
  • Results are achieved rapidly, with the automated analysis of 10 samples performed in approximately 30 minutes
  • Complete data analysis may be performed in one assay, providing sizing, quantitation, and purity information
  • Provides sensitivity comparable to coomassie blue stains across a large linear dynamic range
  • Meet GMP and GLP
En savoir plus