Imprimer

Absolutely total RNA purification kits

Marques : AGILENT
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

400753 400800 400805 400809 400811
AGLS400753EA 438 EUR
AGLS400753 AGLS400800 AGLS400805 AGLS400809 AGLS400811
Absolutely total RNA purification kits
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents

Absolutely RNA Product portfolio makes RNA purification from tissue and cell samples easy and fast.


  • Absolutely Total RNA - efficient spin column purification of RNA available in Nano, Micro, Mini, and 96 well Prep formats
  • Absolutely RNA miRNA Kit - produces high yields of pure miRNA-containing total RNA from cells or tissue samples
  • Absolutely RNA FFPE Kit - delivers a simple and reliable, non-toxic method to isolate quality RNA from paraffin embedded tissue sections


Absolutely RNA miRNA Kit: Designed to produce high yields of pure miRNA containing total RNA from cells or tissue samples, specifically qualified for RT-PCR and QRT-PCR.
Absolutely RNA Miniprep Kit: Designed to produce high yields of pure total RNA from cells or tissue samples, specifically qualified for RT-PCR and QRT-PCR.
Absolutely RNA FFPE Kit: Delivers a simple and reliable, non-toxic method to isolate quality RNA from paraffin embedded tissue sections and includes a quantitative RNA reference standard for meaningful QRT-PCR expression analysis.
Absolutely RNA FFPE Kit (without deparaffinization reagents): Delivers a simple and reliable, non-toxic method to isolate quality RNA from paraffin embedded human and mouse tissue sections and includes a quantitative RNA reference standard for meaningful QRT-PCR expression analysis.
Absolutely RNA 96 uprep Kit: High-throughput RNA purification kit in a 96 well format.
Absolutely RNA Nanoprep Kit: The only kit qualified to isolate concentrated total RNA from the smallest samples, down to a single cell.
Absolutely RNA uprep Kit: Specifically optimised for efficient purification of DNA-free total RNA from cultured cells.


No heating, no long centrifugation, and no repurification is required. Each absolutely RNA purification kit has been optimized for a specific range of sample sizes and elution volumes.


Informations de commande: Absolutely RNA kits include all the reagents you need for DNA-free total RNA, and miRNA isolation with no additional purchase of DNase required.


Attention: For research use only. Not for use in diagnostic procedures.

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable
Application
Target molecule RNA RNA RNA
Refine target molecule Total RNA Total RNA Total RNA
Sample type Human/animal cells and tissues Human/animal cells and tissues FFPE tissues
Method / Format Column (silica-based) Column (silica-based) Column (silica-based)
Prep size Micro Mini Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR