Imprimer
Benchtop cooler, cryo preservation module, StrataCooler, AGLS
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
400005
AGLS400005EA 314 EUR
AGLS400005
Benchtop cooler, cryo preservation module, StrataCooler, AGLS
Refroidisseurs Refroidisseurs de paillasses
Most cell types show >50% survival when cooling rates are kept at 0,2 to 1 °C/min. The StrataCooler Cryo preservation module has been designed to freeze mammalian and insect cells at a controlled rate of 0,4 to 0,6 °C/min. to achieve an 80 to 90% survival rate.
  • 32-(standard) cryovial capacity