Imprimer
BL21 competent cells
Fournisseur: STRATAGENE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
230280 200132 200133 230130 230132 230134 230240 230245 230255 230280 235200
STRA230280EA 0 EUR
STRA230280 AGLS200132 AGLS200133 AGLS230130 AGLS230132 AGLS230134 AGLS230240 AGLS230245 AGLS230255 AGLS230280 AGLS235200
BL21 competent cells
Cellules compétentes
BL21 competent cells are an all-purpose strain for high-level protein expression and easy induction.
  • High-level protein expression and easy induction
  • Can be used with non-T7 RNA polymerase protein expression systems
  • Allows the tightest control of protein expression for extremely toxic proteins
  • Suitable for expressing proteins from all species, including mammals
  • Ideal for difficult protein expression, especially when codon bias is a problem
  • Increases transformation efficiency with Hte phenotype
  • Derived from Agilent’s high performance BL21-Gold competent
En savoir plus