Imprimer
Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid Standard for ICP-MS
Marques : ACTIVE MOTIF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
M208.2NP.L1
ACSDM208.2NP.L1EA 130 EUR
ACSDM208.2NP.L1
Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid Standard for ICP-MS
Étalons Étalons cationiques Standards de bismuth
Bi 1000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
New Product
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable