Imprimer

Anti-RPL17 Rabbit Polyclonal Antibody

Fournisseur: Abgent
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

AP9892B
ABGEAP9892BEA 354 EUR
ABGEAP9892B
Anti-RPL17 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: RPL17 (ribosomal protein L17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR