Imprimer

Anti-GTF2H4 Rabbit Polyclonal Antibody

Fournisseur: Abgent
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

AP8730C
ABGEAP8730CEA 354 EUR
ABGEAP8730C
Anti-GTF2H4 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: GTF2H4 (general transcription factor IIH, polypeptide 4, 52kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR