Imprimer

Anti-MOSPD2 Rabbit Polyclonal Antibody

Fournisseur: Abgent
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

AP13323A
ABGEAP13323AEA 354 EUR
ABGEAP13323A
Anti-MOSPD2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: MOSPD2 (motile sperm domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR