Imprimer

Anti-YDJC Rabbit Polyclonal Antibody

Fournisseur: Abgent
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

AP10640C
ABGEAP10640CEA 354 EUR
ABGEAP10640C
Anti-YDJC Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: YDJC (YdjC homolog (bacterial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR