Imprimer

Hyaluronic Acid (HA) ELISA Kit, Inc.

Marques : Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

AB287799-96
ABCAAB287799-96EA 1160 EUR
ABCAAB287799-96
Hyaluronic Acid (HA) ELISA Kit, Inc.
Tests Tests ELISA

A competitive ELISA kit for the quantitative measurement of hyaluronic acid in biological fluids.


  • Easy, convenient and time-saving method
  • Detection range: 1,563 to 100 ng/ml
  • Sensitivity: <0,938 ng/ml
  • Assay precision: Intra-assay: CV <8%; Inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed


Hyaluronic acid (HA) is an anionic, nonsulfated glycosaminoglycan distributed widely throughout connective, epithelial, and neural tissues. It is unique among glycosaminoglycans in that it is nonsulfated, forms in the plasma membrane instead of the golgi apparatus, and can be very large.


One of the chief components of the extracellular matrix, hyaluronan contributes significantly to cell proliferation and migration, and may also be involved in the progression of some malignant tumors.


Hyaluronic acid ELISA kit is a competitive ELISA assay for the quantitative measurement of hyaluronic acid in serum, plasma and cell culture supernatants.


The density of colour is proportional to the amount of Hyaluronic acid captured from samples.


Attention:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.


Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR