Imprimer

NOS2 / iNOS (Mouse) ELISA Kit

Marques : Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

AB285318-96
ABCAAB285318-96EA 1440 EUR
ABCAAB285318-96
NOS2 / iNOS (Mouse) ELISA Kit
Tests Tests ELISA

A sandwich ELISA kit for the quantitative measurement of NOS2 in biological fluids.


  • Sensitivity: <0,188 ng/ml, detection method absorbance (450 nm)
  • Detection range: 0,312 to 20 ng/ml, assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%, Application- quantitative measurement of NOS2 in serum, plasma, tissue homogenates and other biological fluids
  • No significant cross-reactivity or interference was observed, species reactivity mouse


Nitric oxide synthases (NOS) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. In presence of NADPH, FAD, FMN, (6R)-5,6,7,8-tetrahydrobiopterin, calmodulin and heme, NOS catalyzes a five-electron oxidation of the guanidino nitrogen of L-arginine with molecular oxygen to generate NO and L-citrulline. There are three isoforms of NOS: endothelial (eNOS), neuronal (nNOS), and inducible (iNOS). iNOS is expressed de novo under oxidative stress conditions and produces large amounts of NO as a part of body's defense mechanism. BioVision's NOS2 ELISA kit is a sandwich ELISA assay for the quantitative measurement of NOS2 in mouse serum, plasma and cell culture supernatants. The density of color is proportional to the amount of NOS2 captured from the samples.


Attention:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.


Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR