Imprimer
XFD555 NHS ester [Same Structure to Alexa Fluor™ 555 NHS ester]
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1821 71906
AATB1821EA 309 EUR
AATB1821 AATB71906
XFD555 NHS ester [Same Structure to Alexa Fluor™ 555 NHS ester]
XFD555 NHS ester
XFD555 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 555 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Attention: For Research Use Only