Imprimer
iFluor® 700, Succinimidyl Ester
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1036 71036 71514 71564
AATB1036EA 307 EUR
AATB1036 AATB71036 AATB71514 AATB71564
iFluor® 700, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Attention: For research use only.