Imprimer
MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
22413
AATB22413EA 614 EUR
AATB22413
MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit
Tests Tests immunologiques
The MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit provides an easy and convenient way for determination of gram sign in live bacteria.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimised for fluorescent microscopy
Attention: For research use only.