Imprimer

Calcein AM 95, Ultra Pure Grade

Marques : AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): Calcein-AM, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein acetoxymethyl ester, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate

22003 22004
AATB22003EA 230 EUR
AATB22003 AATB22004
Calcein AM 95, Ultra Pure Grade
Calcein AM

Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein.

Formule: C₄₆H₄₆N₂O₂₃
Poids moléculaire: 994.86 g/mol
Numéro MDL: MFCD05861516
Numéro CAS: 148504-34-1
ADR: 5 II

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR