Imprimer
ReadiLink™ xtra Rapid Cy3 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1971
AATB1971EA 292 EUR
AATB1971
ReadiLink™ xtra Rapid Cy3 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
Systèmes de liaison et de marquage des protéines
ReadiLink™ xtra rapid antibody labelling kits require essentially only 2 simple mixing steps without a column purification needed.
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Attention: For research use only.