Imprimer
ProLite™ 5 to 245 kD Protein Ladder
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
18015
AATB18015EA 133 EUR
AATB18015
ProLite™ 5 to 245 kD Protein Ladder
Réactifs d'électrophorèse Marqueurs de poids moléculaires
The ProLite™ 5 to 245 kD protein ladder is a high-quality three-colour protein standard consisting of 13 prestained proteins.
  • Stable for 12 months
Quality tested in-house.
Attention: For research use only.