Imprimer
Gelite™ Green Nucleic Acid Gel Staining Kit
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
17589
AATB17589EA 307 EUR
AATB17589
Gelite™ Green Nucleic Acid Gel Staining Kit
Tests Tests à l'acide nucléique
Gelite™ Green is a sensitive fluorescent nucleic acid gel stain for detecting nucleic acids in agarose and polyacrylamide gels.
  • 2× more sensitive than etbr
  • Brighter signal
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Attention: For research use only.