Imprimer
PI Brite™
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
17584 17583
AATB17584EA 265 EUR
AATB17584 AATB17583
PI Brite™
PI Brite™
PI Brite™ is a propidium iodide (PI) analog developed by AAT Bioquest to improve the brightness and fixability of propidium iodide in cells.
  • Stable for 24 months
Quality tested in-house
Attention: For Research Use Only