Imprimer

Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy7®)

Fournisseur: AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

16880
AATB16880EA 307 EUR
AATB16880
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy7®)
Anticorps
AAT Bioquest's anti-rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity-purified
  • Bright signal
  • Cross-adsorbed

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR