Imprimer
ReadiLink™ Rapid XFD594 Antibody Labelling Kit 'XFD594 Same Structure to Alexa Fluor™ 594' (2×50 µg Labellings)
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1277
AATB1277EA 218 EUR
AATB1277
ReadiLink™ Rapid XFD594 Antibody Labelling Kit 'XFD594 Same Structure to Alexa Fluor™ 594' (2×50 µg Labellings)
Systèmes de liaison et de marquage des protéines
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Attention: For research use only.