Imprimer

D(+)-Xylose 98

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

A10643.0E A10643.22 A10643.36
A10643.0EEA 865 EUR
A10643.0E A10643.22 A10643.36
D(+)-Xylose 98
D(+)-Xylose

Purity: 98+% CAS No.: 58-86-6 Molecular Formula: C5H10O5 Formula Weight: 150.13 Flash Point: 100°(212°F) Physical Form: Cryst powder Density: 1.525 Sensitivity: Hygroscopic Appearance: White Melting Point: 147-151° Beilstein Reference: 1562108 Merck Reference: 14,10087 Optical Rotation: +19° (c=10 in water + NH4OH) MDL No.: MFCD00151475

Formule: C₅H₁₀O₅
Poids moléculaire: 150.13 g/mol
Point d'ébullition: 331
Point de fusion: 147…151 °C
Densité: 1.525
Point éclair: >100 °C (212 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00151475
Numéro CAS: 58-86-6
EINECS: 200-400-7
Indice Merck: 13,10142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR