Imprimer

Chloroforme 99,8 GC, corrected for water content Stabilised, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®

Marques : Avantor
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Trichlorométhane

Stabilisation: ~0.75% EtOH
Inert_Gas_filled

9257-03EA 376.78 EUR
9257-03 9257-02
Chloroforme 99,8 GC, corrected for water content Stabilised, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®
Chloroforme
Formule: CHCl₃
Poids moléculaire: 119.38 g/mol
Point d'ébullition: 61.7
Point de fusion: –63 °C
Densité: 1.4832 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00000826
Numéro CAS: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Indice Merck: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

For Laboratory,Research,or Manufacturing Use = 10
Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤10
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Chloride (Cl) ≤10 ppm
Color (APHA) ≤10
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Titrable Acid (µeq/g) ≤0.5
Water (by KF, coulometric) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR