Imprimer
Small ligand labelling kits, Mix-n-Stain™ CF® Dye
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
92350 92351 92352 92353 92354 92355 92356 92357 92358 92359 92360 92361 92362 92363 92364
92350.EA 293 EUR
92350. 92351. 92352. 92353. 92354. 92355. 92356. 92357. 92358. 92359. 92360. 92361. 92362. 92363. 92364.
Small ligand labelling kits, Mix-n-Stain™ CF® Dye
Systèmes de liaison et de marquage des protéines
Easily label SNAP-tag®, CLIP-tag™, HaloTag®, or other ligands with fluorescent dyes, no purification needed. Cell membrane-permeant and impermeant dye options for intracellular or surface targets.
  • For SNAP-tag®, CLIP-tag™ and HaloTag®, or other ligands with an aliphatic amine
  • Label 100 nmol ligand
  • Reaction takes ~30 minutes with minimal hands-on time and no purification
  • Choose from 15 bright and photostable CF® Dyes
  • Cell membrane-permeant or impermeant options for intracellular or surface targets
Mix-n-Stain™ small ligand labelling kits are designed for rapid labeling of low molecular weight (MW ~150 to 5000), high affinity biological ligands or substrates.
Biotium also offers TMP-PEG3-Amine for making fluorescent TMP ligands. Also see Biotium's Mix-n-Stain™ antibody labelling kits available with bright and photostable CF® Dyes and other labels.
SNAP-tag and CLIP-tag are trademarks or registered trademarks of New England BioLabs; HaloTag is a registered trademark of Promega Corporation.