Imprimer
Supelco® TraceCERT® Organic Reference Standard, Atrazine
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
90935-50MG
90935-50MGEA 75.7 EUR
90935-50MG
Supelco® TraceCERT® Organic Reference Standard, Atrazine
Étalons Étalons organiques
  • GHS signal: GHS08, GHS07, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Attention: P260 - P272 - P273 - P280 - P302 + P352 - P314