Imprimer
Software for HPLC method development, ChromSwordAuto® 5 and ChromSwordAuto® 5 Hitachi Edition
Fournisseur: COLLECTION PREFIX HPLC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
908-0062 908-0063 908-0064 908-0065 908-0066 908-0080 908-0081 908-0082 908-0083 908-0084 908-0085 908-0086 908-0087 908-0088 908-0090 908-0091 908-0092 908-0093 908-0094 908-0095 908-0096 908-0097
908-0062EA 16320 EUR
908-0062 908-0063 908-0064 908-0065 908-0066 908-0080 908-0081 908-0082 908-0083 908-0084 908-0085 908-0086 908-0087 908-0088 908-0090 908-0091 908-0092 908-0093 908-0094 908-0095 908-0096 908-0097
Software for HPLC method development, ChromSwordAuto® 5 and ChromSwordAuto® 5 Hitachi Edition
Logiciels Chromatography Software
ChromSwordAuto® 5: Communicates directly with the HPLC instrument in order to create a fully automated HPLC or UHPLC method development station. The unique ChromSwordAuto® software is capable of developing and optimising HPLC methods fully automatically within just a few hours. Methods already in use can also be optimised, thus reducing analysis time considerably. All you have to do is place the sample in the autosampler, set a few parameters and start the optimisation process. ChromSwordAuto® then predicts the method conditions and controls the HPLC system connected in recording theEn savoir plus