Imprimer
LC-MS/MS system, SCIEX Triple Quad 3500
Fournisseur: AB SCIEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5062231 5073097 5073096 5073675 5073676 5078671 5078672 5078678 5078634 5078635 5078636 5078681 5087706 5078640 5078673 5078674 5078675 5078642 5078644 5078646 5078638 5078639
904-0020EA 149680 EUR
904-0020 904-0021 904-0022 904-0023 904-0024 904-0036 904-0037 904-0038 904-0039 904-0040 904-0041 904-0044 904-0045 904-0046 904-0047 904-0048 904-0049 904-0050 904-0051 904-0052 904-0042 904-0043
LC-MS/MS system, SCIEX Triple Quad 3500
Spectrophotomètres et Photomètres Mass Spectrometers
The 3500 Triple Quad system offers productivity, reliability, and robustness in a modernised entry-level mass spec for today’s analytical laboratories. The Triple Quad 3500 system offers the speed and precision you want from a modern mass spec system, with the legendary performance and dependability you trust from SCIEX technology.
  • Qurved LINAC® collision cell: More MRMs per injection, space-saving design
  • High pressure Q0 and QJet® ion guide: Enhanced sensitivity through more efficient ion focusing
  • Fast eQ™ electronics: Compatible with fast LC — achieve
En savoir plus