Imprimer
Filling nozzle clamps
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
9.67106.0001 9.67107.0001
9.67106.0001EA 225 EUR
9.67106.0001 9.67107.0001
Filling nozzle clamps
Pinces Pinces à usage général
Stainless steel clamps for filling nozzles, with drip-free attachment to drums or to walls.
Safety and convenient handling are the basis for an important part of very special Merck product developments. When using a pressurizing withdrawal system for stainless steel drums the Merck filling nozzle (9.67100.1090 / 9.67100.9090) can be placed safely even directly on the small rim of the drum by using our screw clamp. For individual solvent withdrawal installations that end with a flexible filling nozzle inside a fume hood, a clamp for wall fastening is now available.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable