Imprimer
TISAB I Solution, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
89465-500ML
89465-500MLEA 123 EUR
89465-500ML
TISAB I Solution, Supelco®
Tampons Tampons d'ajustement de la force ionique
TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes.
TISAB I can be prepared using NaCl with glacial acetic acid, tri-sodium citrate and NaOH.
TISAB I has been used in determination of fluoride-ion concentration using fluoride ion sensitive electrode.