Imprimer

Alcool benzylique 99,5, PESTINORM® GC_HS

Marques : VWR Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): Phénylcarbinol

88250.290EA 175 EUR
88250.290
Alcool benzylique 99,5, PESTINORM® GC_HS
Alcool benzylique
Formule: C₆H₅CH₂OH
Poids moléculaire: 108.14 g/mol
Point d'ébullition: 205 °C (1013 hPa)
Point de fusion: –15,3 °C
Densité: 1.05 g/cm³ (20 °C)
Point éclair: 101 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00004599
Numéro CAS: 100-51-6
EINECS: 202-859-9
Indice Merck: 13,01126

Commander maintenant

Résultats des tests de spécification

Purity (GC, on anhydrous basis) 99.5-100 %
Identity (IR) Complying
Residue after evaporation 0-0.0003 %w/w
Water (KF) 0-0.1 %w/w
Color (APHA, Pt-Co) 0-10
GC-HS (Each class 1 solvent) 0-1 ug/g
GC-HS (Each class 2 solvent) 0-10 ug/g
GC-HS (Each class 3 solvent) 0-50 ug/g
Benzaldehyde content (on botteling) 0-0.01 %
GC-HS (Each unknown) 0-10 ug/g
Acidity 0-0.001 meq/gr
Alkalinity 0-0.002 meq/gr
Suitable as solvent for the analysis of residual solvents of ICH classes 1, 2 and 3 according to Ph Eur and USP.
Each unknown measured against butyl methyl ketone up to anisole is ≤ 10 µg/g.
Formic acid, acetic acid, dimethylacetamide and dimethyl sulfoxide are not detected.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR