Imprimer

VWR® ARISTAR®, Osmium Standard Solution for ICP

Marques : VWR Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

87051.180EA 540 EUR
87051.180
VWR® ARISTAR®, Osmium Standard Solution for ICP
Étalons Étalons cationiques Standards d'osmium

  • Produced from high purity OsCl₃
  • Certified reference material
  • Traceable to NIST
  • Certified Value and uncertainty 1000,0 ± 0,5 mg/L


Density 1,060 g/cm³ at 20 °C.


Attention: Storage at ambient temperatures.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR