Imprimer
Chlorine, sensitive test strips, Dosatest
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
85712.601EA 38 EUR
85712.601
Chlorine, sensitive test strips, Dosatest
Kits de tests environnementaux
Semi-quantitative test strips for the determination of chlorine.
  • Ideal for rapid and easy analysis of chlorine directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to violet
Informations de livraison: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box