Imprimer
Copper test strips, Dosatest
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
85696.601EA 33 EUR
85696.601
Copper test strips, Dosatest
Kits de tests environnementaux
Semi-quantitative test strips for the determination of copper.
  • Ideal for rapid and easy analysis of copper directly on-site
  • No maintenance or calibration is required
Colour change: White to red-violet.
Informations de livraison: Delivered as 100 test strips in a tube with instruction leaflet.
Attention: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.