Imprimer
ESC (Easy Surface Checking) swabs
Fournisseur: LIOFILCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
85607 85601 85602 85603 85604 85605 85606 85608 85609 85610 85612 85614 85615 85621 85623
85607.EA 224 EUR
85607. 85601. 85602. 85603. 85604. 85605. 85606. 85608. 85609. 85610. 85612. 85614. 85615. 85621. 85623.
ESC (Easy Surface Checking) swabs
Systèmes de surveillance Tests de contrôle de l'hygiène
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
  • Pre-filled swab device
  • Rayon bud
  • Wide range of diluents
  • Leak proof closure
  • Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).