Imprimer

n-Heptane 99, HiPerSolv CHROMANORM® LC-MS

Marques : VWR Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): high purity solvents

Nitrogen_filled

84899.320EA 187 EUR
84899.320 84899.290
n-Heptane 99, HiPerSolv CHROMANORM® LC-MS
n-Heptane
Formule: H₃C(CH₂)₅CH₃
Poids moléculaire: 100.20 g/mol
Point d'ébullition: 98.4 °C (1013 hPa)
Point de fusion: –90,6 °C
Point éclair: -4
Numéro MDL: MFCD00009544
Numéro CAS: 142-82-5
UN: 1206
ADR: 3 II
Indice Merck: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Assay (on anhydrous substance) ≥ 99 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Evaporation residue ≤ 1 ppm
Water ≤ 50 ppm
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 25 ppb
Ca (Calcium) ≤ 25 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 10 ppb
K (Potassium) ≤ 10 ppb
Mg (Magnesium) ≤ 10 ppb
Mn (Manganese) ≤ 10 ppb
Na (Sodium) ≤ 50 ppb
Ni (Nickel) ≤ 10 ppb
Pb (Lead) ≤ 10 ppb
Zn (Zinc) ≤ 10 ppb
Transmittance (195 nm) ≥ 10 %
Transmittance (210 nm) ≥ 50 %
Transmittance (220 nm) ≥ 80 %
Transmittance (245 nm) ≥ 95 %
Transmittance (290 nm) ≥ 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR