Imprimer

n-Pentane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Marques : Avantor
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Pentane

8114.2500EA 258.89 EUR
8114.2500 8114.1000 8114.9025 8114.9200
n-Pentane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
n-Pentane
Formule: H₃C(CH₂)₃CH₃
Poids moléculaire: 72.15 g/mol
Point d'ébullition: 35…36 °C (1013 hPa)
Point de fusion: –130 °C
Densité: 0.63 g/cm³ (20 °C)
Point éclair: –40 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00009498
Numéro CAS: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Indice Merck: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Assay (by GC) min. 99 %
Boiling Range 35 - 37 °C
Density (g/ml) at 20°C 0.624 - 0.626
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.005 %
Water (H2O) max. 0.01 %
Trace Impurities (in ppm)Aluminium (Al) max. 0.5
Trace Impurities (in ppm)Boron (B) max. 0.02
Trace Impurities (in ppm)Barium (Ba) max. 0.1
Trace Impurities (in ppm)Calcium (Ca) max. 0.5
Trace Impurities (in ppm)Cadmium (Cd) max. 0.05
Trace Impurities (in ppm)Cobalt (Co) max. 0.02
Trace Impurities (in ppm)Chromium (Cr) max. 0.02
Trace Impurities (in ppm)Copper (Cu) max. 0.02
Trace Impurities (in ppm)Iron (Fe) max. 0.1
Trace Impurities (in ppm)Magnesium (Mg) max. 0.1
Trace Impurities (in ppm)Manganese (Mn) max. 0.02
Trace Impurities (in ppm)Nickel (Ni) max. 0.02
Trace Impurities (in ppm)Lead (Pb) max. 0.1
Trace Impurities (in ppm)Tin (Sn) max. 0.1
Trace Impurities (in ppm)Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR