Imprimer

Eau, molecular grade DNAse_RNAse_free

Marques : G Biosciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): Hydrogen oxide

786-72C 786-73C 786-292 786-293
786-72CEA 221 EUR
786-72C 786-73C 786-292 786-293
Eau, molecular grade DNAse_RNAse_free
Eau

Molecular Grade Water is ultrapure water that certified RNase, DNase and Protease free, is suitable for use in molecular biology applications, which demand a high quality of water and assurance that the water is free from nucleases and protease contamination. No toxic agents, such as DEPC, are used in the manufacturing of Molecular Grade Water, eliminating DEPC interference of enzymatic reactions.


  • Each lot is quality tested for the absence of RNase, DNase and protease contamination


For use in molecular biology applications, PCR reactions, 2D electrophoresis and mass spectrometry.

Formule: H₂O
Poids moléculaire: 18.02 g/mol
Point d'ébullition: 100
Point de fusion: 0
Température de stockage: Température ambiante
Numéro MDL: MFCD00011332
Numéro CAS: 7732-18-5
Indice Merck: 13,10098

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR