Imprimer
Protein purification kit, STRATEGY™
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
786-184A 786-184C 786-184HP
786-184AEA 282 EUR
786-184A 786-184C 786-184HP
Protein purification kit, STRATEGY™
Protéines purification Protein Purification Kits
STRATEGY™ is designed to allow researchers to rapidly screen a number of purification techniques in a small scale format to identify the system to isolate their protein of interest. The self-contained kit allows researchers to save time and money as separate buffers, resins and columns are not needed.
The kit guides the researcher step-by-step through various protein purification techniques like pH fractionation, salt fractionation, hydrophobic chromatography, anionic chromatography and cationic chromatography.