Imprimer
Protein fractionation kit, FOCUS™ mitochondria
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
786-022
786-022EA 689 EUR
786-022
Protein fractionation kit, FOCUS™ mitochondria
Protéines extraction Protein Extraction Kits
FOCUS™ Mitochondria kit is specifically designed for the isolation of intact mitochondria from cultured mammalian cells. This kit allows for the fast and efficient fractionation of the cytoplasm of cultured mammalian cells into an enriched fraction of mitochondria. The majority (>90%) of the mitochondria have intact inner and outer membranes and therefore retain their functionality.
  • Fast and efficient fractionation of active heavy and light mitochondria
  • Suitable for cultured animal cells and tissues
The FOCUS™ Mitochondria kit is highly adaptable for use with animal tissues and other sources of mitochondria. The kit is designed for 50 preps, where one prep is equivalent to 2×10⁷ mammalian cells or 100 mg mammalian tissue.