Imprimer
Filter plates, 24-well, AcroPrep™
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
97026 97016 97017 97027 97047 97048 97049 97050 97051 97052 97053 97054 97055 97056 97057 97058 97059 97060 97061 97062 97029 97030 97031 97032 97033 97034 97035 97036
738-0219EA 736 EUR
738-0219 738-0220 738-0222 738-0221 738-0238 738-0237 738-0236 738-0235 738-0234 738-0233 738-0232 738-0231 738-0230 738-0229 738-0228 738-0227 738-0226 738-0225 738-0224 738-0223 738-0246 738-0245 738-0244 738-0243 738-0242 738-0241 738-0240 738-0239
Filter plates, 24-well, AcroPrep™
Microplaques Plaques de filtration
The Acroprep 24-well filter plate utilises Pall’s proprietary high performance multi-layer filter media and membranes to offer time savings, strong performance claims and streamlined workflow improvements in a 24-well plate format.
  • Intrinsic Acroprep plate and membrane properties minimise sample loss from non specific binding
  • Dual-layer clarification plate contains a depth filter plus 0,65/0,2 μm Supor EKV PES membrane, and allows clarification of large cellular debris prior to filtration through the 0,65/0,2 μm EKV Supor membrane
  • Dual-layer clarification
En savoir plus