Imprimer
Microplates, 96-well, non binding
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
650901_40 655901_40 655900_40 655906_40 655903_40 655904_40 651901_40
738-0165EA 319 EUR
738-0165 738-0170 738-0169 738-0173 738-0171 738-0172 738-0166
Microplates, 96-well, non binding
Microplaques Microplaques HTS
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.