Imprimer
Tubes immunologie Greiner
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
115001_2000 115061_2000 115070_2000 115071_2000
738-0000EA 215 EUR
738-0000 738-0001 738-0002 738-0003
Tubes immunologie Greiner
Tubes Tubes pour immunologie
PS, clear. Star bottomed tubes have an increased surface area and thus make it possible to bind larger amounts of antigens or antibodies.
Immuno tubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone).
Available with either MICROLON® 200 (medium binding) and MICROLON® 600 (high binding) surfaces.
Ø×L: 12×75 mm