Imprimer
Optical bottom plates, SCREENSTAR
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
655866_16 789866_68
735-0387EA 724 EUR
735-0387 735-0389
Optical bottom plates, SCREENSTAR
Microplaques Microplaques HTS
Cycloolefin, black pigmented frame, 190 μm ultra-clear film bottom, tissue culture treated, sterile. SCREENSTAR microplates are specialised products for sophisticated microscopic applications, like high content screening (HCS) or high resolution microscopy with water and oil immersion objectives.
  • Resistant against polar solvents (DMSO)
  • Glass-like optical qualities with a refractive index similar to glass
  • Ideal surface for adherent cell culture
  • Universal microscope objective compatibility
Manufactured entirely of cycloolefin, the microplates feature aEn savoir plus