Imprimer
VWR® Single Use 3D Tank Liners with Bottom Drain
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
734-3361EA 0 EUR
734-3361 734-3362 734-3363 734-3364 734-3365
VWR® Single Use 3D Tank Liners with Bottom Drain
Sachets Sachets pour milieux de culture
These tank liners are single-use bags with an open top, designed to fit in cylindrical vessels.
  • No cleaning after media preparation
  • Medical grade, multi-layer film
  • For bioprocess applications
  • Delivered gamma irradiated and ready-to-use
These bags which have a bottom drain are used as a liner into a vessel to avoid cleaning of the vessel after media and buffer preparation. After installing the Tank Liner into a vessel and filling it with liquid, powder can be added to the vessel and the mixing process can start by using an overhead agitator. WhenEn savoir plus