Imprimer
Racks, magnetic separation, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
F199000000 F199000001 F199000002 F199000003 F199000004
734-2889EA 1350 EUR
734-2889 734-2890 734-2891 734-2892 734-2893
Racks, magnetic separation, SP Bel-Art
Portoirs Portoirs de séparation magnétique
Acrylic, clear.
  • Independent sleeves make liquid addition and removal easier with less accidental fluid loss
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves allows beads to be sequestered at various heights along the tube wall
  • Simply turn tube for efficient bead mixing
These magnetic separation racks has sequestered magnetic beads, so that rinse solutions and affinity ligands such as antibodies, streptavidin, or proprietary reagents for coupling nucleic acids can be added or decanted in assays. The 96-well PCR plate model has 24En savoir plus