Imprimer
Amine cell culture flasks, Corning® PureCoat™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
354726 354728 356726 356728
734-2212EA 72.9 EUR
734-2212 734-2266 734-2268 734-2407
Amine cell culture flasks, Corning® PureCoat™
Ballons Flacons pour culture cellulaire
PS, with blue screw cap.
Corning® PureCoat™ amine flasks provide an enhanced tissue culture surface. The positively charged surface, manufactured using a proprietary thin-film coating technology, offers improved attachment, increased proliferation, and enhanced recovery from freeze-thaw for a broad range of primary, transfected and fastidious cells that exhibit poor attachment properties on standard tissue culture (TC) surfaces in serum-free/serum-reduced conditions.
Attention: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment,En savoir plus