Imprimer
Fibronectin-coated flasks, Corning® BioCoat®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
354521 354526
734-0180EA 498 EUR
734-0180 734-0184
Fibronectin-coated flasks, Corning® BioCoat®
Ballons Flacons pour culture cellulaire
Human Fibronectin (HFN) is a widely distributed glycoprotein that is used as a substrate to promote attachment of cells through its central-binding domain RGD sequence. HFN is a product of most mesenchymal and epithelial cells and is present in both the ECM and plasma. The principal function of HFN appears to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis
Applications include promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cellEn savoir plus