Imprimer
sparQ PureMag beads
Marques : Quantabio
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
95196-450 95196-060 95196-005
733-2628EA 4030 EUR
733-2628 733-2627 733-2626
sparQ PureMag beads
Réactifs pour acides nucléiques Sets de réactifs pour séquençage de nouvelle génération
sparQ PureMag beads is a fast and reliable nucleic acid purification system for reaction cleanup and size selection in Next Generation Sequencing (NGS) workflows. Based on the reversible nucleic acid-binding properties of magnetic beads; this product can be used to quickly remove primers, primer-dimers, unincorporated nucleotides, salts, adapters and adapter-dimers from NGS library prep reactions to improve downstream sequencing performance. sparQ PureMag beads allows excellent recovery of fragments greater than 100 bp without centrifugation or filtration. Consistent and reliable size selection can be achieved by simply adjusting the beads to sample ratio. This product is designed for both manual and automated processing, allowing seamless integration into existing workflows.
  • High recovery of DNA fragments greater than 100 bp
  • Efficient removal of unwanted components from adapter ligation and PCR reactions
  • Consistent single- or double-sided size selection
  • Seamless integration into existing NGS workflows with little or no protocol change
  • Easy to use and compatible with manual processing or automated liquid handling robots
  • Cost effective alternative to AMPure® XP with equivalent performance