Imprimer

Gel documentation system, Syngene G:BOX Chemi XX6

Fournisseur: Syngene (a division of the Synoptics group)
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

GBOX-CHEMI-XX6
733-2351EA 32530 EUR
733-2351
Gel documentation system, Syngene G:BOX Chemi XX6
Systèmes de photodocumentation
The Syngene G:BOX Chemi XX6 gel documentation system offers optimum performance. Highly sensitive, it is designed for advanced chemiluminescent Western blots, multiplex gels, IR and routine imaging of fluorescent and visible gels and blots. The higher spatial resolution of this system makes it suitable for advanced applications, including 2D. The G:BOX Chemi XX6 includes a super-high resolution, 6 megapixel camera with quantum efficiency >73% at 425 nm, giving outstanding sensitivity for chemiluminescence. Functional control of the G:BOX Chemi XT4 is handled by the GeneSysEn savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR