Imprimer
VWR®, PCR/qPCR Plates, 96-Wells, Plastic
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
732-3753EA 160 EUR
732-3753 732-3754 732-3755 732-3756 732-3757 732-3758 732-3759 732-3760 732-3761 732-3762 732-3763 732-3764 732-3765 732-3766 732-3767 732-3768 732-3769 732-3770
VWR®, PCR/qPCR Plates, 96-Wells, Plastic
Microplaques Plaques PCR
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results.
  • Natural or white PP
  • Standard or low profile
  • Flat top or raised deck
  • Non skirted, semi-skirted or fully skirted
  • Options to fit standard, ABI FAST and Roche 96-well blocks
  • Autoclavable
96-well PCR plates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.
Certifications: FDA 21 CFR compliance. Compliance to BSE/TSE (free from animal origin). Non pyrogenic, DNase-, RNase- and latex-free.
Informations deEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable